Learn More
Invitrogen™ CytoVista™ Tissue Imaging Chamber, 0.75 mm deep
Description
These chambers are made of silicon and are chemically inert, and so are resistant to any clearing reagent. After fluorescently labeling and clearing tissue or 3D cell culture, place the specimen in this chamber along with imaging solution. Then place a coverslip on top and you are ready to image.
Features of the CytoVista family of tissue and 3D cell culture clearing reagents include:
• Rapid clearing of fluorescently labeled tissue for 3D imaging
• Minimal shrinkage, expansion, or other morphological changes to tissue
• Simple workflow does not require any special instruments or equipment and is easily adapted to IHC
• Compatible with most fluorophores, including dyes and fluorescent proteins
The CytoVista tissue and 3D cell culture clearing workflow clears tissue or 3D cultured cells with minimal changes to morphology or the sensitivity of detection with almost any fluorophore. With an easy-to-use protocol, whole mouse brain (8-mm section) can be cleared in 24 hours, or a 1-mm section in 2 hours, without the use of any special instruments or equipment.
The CytoVista clearing workflow is compatible with most fluorophores, including fluorescent proteins that are detected with common fluorescent imaging instruments, such as wide-field microscopes, high content analyzers, confocal microscopes, and lightsheet microscopes. For first time users, we recommend the CytoVista Tissue Clearing/Staining Kit (Cat No V11324) or CytoVista 3D Cell Culture Clearing/Staining Kit (Cat No. V11325). These kits contain all necessary reagents for both clearing and IHC staining.
CytoVista clearing technology sufficiently clears tissue for 3D fluorescent imaging, but unlike other clearing techniques does not significantly changes the tissue morphology. Minimal morphological changes, such as shrinkage and contraction, have been observed. For precious samples, the clearing can be reversed, and the tissue can be processed for histology studies, like H&E staining.
For tissues that are easier to clear, like brain, lung, intestine, skin, and tissues of less than 250-μm thickness, treatment with CytoVista Tissue Clearing Reagent is sufficient for 3D imaging. However, tissues of more than 250-μm thickness or that are hard to clear like kidney, liver, heart, and placenta will require an initial treatment with CytoVista Tissue Clearing Reagent followed by treatment with CytoVista Tissue Clearing Enhancer.
CytoVista 3D Cell Culture Clearing Reagent is recommended for spheroids or organoids up to 1 mm in thickness.
For hard tissues like bone, cartilage, and connective tissue, CytoVista Tissue Clearing Reagent is not recommended.
Specifications
Specifications
| Type | Imaging Chamber |
| Surface Treatment | Cell Culture |
| Material | Silicone |
| Thickness (Metric) | 0.75 mm |
| Content And Storage | Store at room temperature. |
| Shipping Condition | Room Temperature |
| No. per Pack | 6 chambers |
| Product Line | Image-iT |
| Quantity | 6 chambers |
Frequently Asked Questions (FAQs)
This should not be required as the silicone naturally adheres to the glass. If sealing is desired, you may seal with melted paraffin or VALAP at the outer edges.
The CytoVista Tissue Imaging chambers have an outer perimeter of 25 mm x 25 mm and an inner diameter of 16 mm; they vary in thickness.
The silicone naturally adheres to glass.
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.