Learn More
Aviva Systems Biology Corp Cyyr1 Antibody - middle region (ARP45884_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP45884_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Cyyr1. It was validated on Western Blot. The immunogen is a synthetic peptide corresponding to a region of Rat. Alternate names: Cyyr1ApplicationWB. Species reactivity: Dog, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-285-225
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.