Learn More
Medchemexpress LLC HY-100222 1mg Medchemexpress, CZ415 CAS:1429639-50-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1002221MG
Encompass_Preferred
Medchemexpress, HY-100222 1mg CZ415 CAS:1429639-50-8 CZ415 is a potent and highly selective mTOR inhibitor with a pIC 50 of 8.07. CZ415 inhibits mTORC1 and mTORC2 protein complex. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-1760