Learn More
Bioworld Czapek Dox Agar, 100 g
Catalog No. 50254412
Encompass_Preferred
Czapek Dox AgarUsed for fungi and bacteria that are unable to utilize sodium nitrate a their sole source of nitrogenWorking Concentration: 50.1 g/LpH: 7.3+/-0.2Dehydrated Appearance: off-white to beige powderPrepared Appearance: colorless agarSoluble in boiling waterContains:SaccharoseSodium NitrateDipotassium PhosphateMagnesium SulfatePotassium ChlorideFerrous SulfateAgarUsage : bioWORLD's products are supplied for LABORATORY RESEARCH USE ONLY. The product may not be used as a drug, agricultural or pesticidal product , food additive or as a household chemical.
Catalog No. 50-254-412 Supplier Bioworld Supplier No. 306201111
Czapek Dox AgarUsed for fungi and bacteria that are unable to utilize sodium nitrate a their sole source of nitrogenWorking Concentration: 50.1 g/LpH: 7.3+/-0.2Dehydrated Appearance: off-white to beige powderPrepared Appearance: colorless agarSoluble in boiling waterContains:SaccharoseSodium NitrateDipotassium PhosphateMagnesium SulfatePotassium ChlorideFerrous SulfateAgarUsage : bioWORLD's products are supplied for LABORATORY RESEARCH USE ONLY. The product may not be used as a drug, agricultural or pesticidal product , food additive or as a household chemical.