Learn More
MOLTOX Molecular Toxicology Inc Czapek Solution Agar, 25 mL/plate, 10 plates/sleeve, Store at Room Temperature
SDP
Supplier:  MOLTOX Molecular Toxicology Inc 2140S133
Encompass_Preferred
Used for the identification of Aspergillus, Penicillium and Paecilomyces as recommended in Standard Methods for the Examination of Water and Wastewater. Each lot of media from MOLTOX is tested for sterility, where applicable, and its ability to support the growth of the appropriate microorganism. All media are accompanied by GLP-compliant formulation and Quality Control certificate.
Catalog No. 50-948-628