Learn More
TARGETMOL CHEMICALS INC CZC-25146 10MG
SDP
Catalog No. 502577013
Encompass_Preferred
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. CZC-25146 (CHEMBL2397014) is an effective specific and metabolically stable LRRK2 inhibitor with IC50 of 4.76/6.87 nM for wild-type LRRK2(Leucine-rich repeat kinase-2) and G2019S LRRK2 respectively. purity: 98%
Catalog No. 50-257-7013 Supplier TARGETMOL CHEMICALS INC Supplier No. T305310MG
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. CZC-25146 (CHEMBL2397014) is an effective specific and metabolically stable LRRK2 inhibitor with IC50 of 4.76/6.87 nM for wild-type LRRK2(Leucine-rich repeat kinase-2) and G2019S LRRK2 respectively. purity: 98%