Learn More
Selleck Chemical LLC CZC-54252
SDP
Catalog No. 5000002921
Encompass_Preferred
CZC-54252 is a potent and selective LRRK2 inhibitor with IC50s of 1 28 nM and 1 85 nM for human wild type LRRK2 and G2019S LRRK2 respectively
Catalog No. 50-000-02921 Supplier Selleck Chemical LLC Supplier No. S65345MG
CZC-54252 is a potent and selective LRRK2 inhibitor with IC50s of 1 28 nM and 1 85 nM for human wild type LRRK2 and G2019S LRRK2 respectively