Learn More
Medchemexpress LLC Czc-54252 hydrochloride | 1784253-05-9 | 98.4% | 1 ML
SDP
Catalog No. 5000409321
Encompass_Preferred
CZC-54252 hydrochloride is a potent and selective LRRK2 inhibitor with IC50 values of 1.28 nM for wild-type LRRK2 and 1.85 nM for G2019S LRRK2. It attenuates G2019S LRRK2-induced human neuronal injury with an EC50 of approximately 1 nM and demonstrates neuroprotective activity. This compound is intended for research use only.
  • Potent and selective LRRK2 inhibitor
  • Attenuates G2019S LRRK2-induced human neuronal injury
  • Demonstrates neuroprotective activity
  • For research use only
Catalog No. 50-004-09321 Supplier Medchemexpress LLC Supplier No. HYB0792A10MM/1ML
CZC-54252 hydrochloride is a potent and selective LRRK2 inhibitor with IC50 values of 1.28 nM for wild-type LRRK2 and 1.85 nM for G2019S LRRK2. It attenuates G2019S LRRK2-induced human neuronal injury with an EC50 of approximately 1 nM and demonstrates neuroprotective activity. This compound is intended for research use only.
  • Potent and selective LRRK2 inhibitor
  • Attenuates G2019S LRRK2-induced human neuronal injury
  • Demonstrates neuroprotective activity
  • For research use only