Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC CZC-54252 (hydrochloride) | 1784253-05-9 | 98.4% | 25 MG
SDP

Catalog No. 5000450453
Encompass_Preferred

CZC-54252 hydrochloride is a potent and selective LRRK2 inhibitor. It attenuates G2019S LRRK2-induced human neuronal injury and exhibits neuroprotective activity. This product is a solid, typically light yellow to orange in appearance.

  • Potent and selective LRRK2 inhibitor
  • Attenuates G2019S LRRK2-induced human neuronal injury
  • Exhibits neuroprotective activity
  • Molecular weight: 541.45
  • Formula: C22H26Cl2N6O4S
  • Appearance: Solid, light yellow to orange
  • Stable under recommended storage conditions
  • Soluble in DMSO

Catalog No. 50-004-50453 Supplier Medchemexpress LLC Supplier No. 25MGHYB0792A
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CZC-54252 hydrochloride is a potent and selective LRRK2 inhibitor. It attenuates G2019S LRRK2-induced human neuronal injury and exhibits neuroprotective activity. This product is a solid, typically light yellow to orange in appearance.

  • Potent and selective LRRK2 inhibitor
  • Attenuates G2019S LRRK2-induced human neuronal injury
  • Exhibits neuroprotective activity
  • Molecular weight: 541.45
  • Formula: C22H26Cl2N6O4S
  • Appearance: Solid, light yellow to orange
  • Stable under recommended storage conditions
  • Soluble in DMSO

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.