Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC CZC-54252 hydrochloride | 1784253-05-9 | 98.4% | 5 MG
SDP

Catalog No. 5000407735
Encompass_Preferred

CZC-54252 hydrochloride is a potent and selective LRRK2 inhibitor with IC50 values of 1.28 nM for wild-type LRRK2 and 1.85 nM for G2019S LRRK2. This compound can attenuate G2019S LRRK2-induced human neuronal injury with an EC50 of approximately 1 nM, exhibiting neuroprotective activity.

  • Potent and selective LRRK2 inhibitor
  • Attenuates G2019S LRRK2-induced human neuronal injury
  • Exhibits neuroprotective activity
  • Molecular weight: 541.45
  • Chemical formula: C22H26Cl2N6O4S
  • Appearance: Solid
  • Color: Light yellow to orange
  • Recommended storage: 4°C, sealed, away from moisture

Catalog No. 50-004-07735 Supplier Medchemexpress LLC Supplier No. HYB0792A5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CZC-54252 hydrochloride is a potent and selective LRRK2 inhibitor with IC50 values of 1.28 nM for wild-type LRRK2 and 1.85 nM for G2019S LRRK2. This compound can attenuate G2019S LRRK2-induced human neuronal injury with an EC50 of approximately 1 nM, exhibiting neuroprotective activity.

  • Potent and selective LRRK2 inhibitor
  • Attenuates G2019S LRRK2-induced human neuronal injury
  • Exhibits neuroprotective activity
  • Molecular weight: 541.45
  • Chemical formula: C22H26Cl2N6O4S
  • Appearance: Solid
  • Color: Light yellow to orange
  • Recommended storage: 4°C, sealed, away from moisture

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.