Learn More
Medchemexpress LLC CZC-8004 | 916603-07-1 | 99.8% | 309.34 | 1 ML
SDP
Catalog No. 5000343088
Encompass_Preferred
CZC-8004 is a pan-kinase inhibitor that demonstrates potent activity against EGFR WT and VEGFR2 V916M. It is utilized in cancer research, particularly in studies involving various human cancer cell lines.
  • Pan-kinase inhibitor
  • IC50 values of 650 nM for EGFR WT
  • IC50 values of 437 nM for VEGFR2 V916M
  • Used in cancer research
  • Targeting ability to H1975 cells with VEGFR2 V916M mutation
  • White to off-white solid appearance
  • Soluble in DMSO
Catalog No. 50-003-43088 Supplier Medchemexpress LLC Supplier No. HY111138100MG
CZC-8004 is a pan-kinase inhibitor that demonstrates potent activity against EGFR WT and VEGFR2 V916M. It is utilized in cancer research, particularly in studies involving various human cancer cell lines.
  • Pan-kinase inhibitor
  • IC50 values of 650 nM for EGFR WT
  • IC50 values of 437 nM for VEGFR2 V916M
  • Used in cancer research
  • Targeting ability to H1975 cells with VEGFR2 V916M mutation
  • White to off-white solid appearance
  • Soluble in DMSO