Learn More
Medchemexpress LLC CZC-8004 | 916603-07-1 | 99.8% | 309.34 | 10 MG
SDP
Catalog No. 5000344178
Encompass_Preferred
CZC-8004 is a pan-kinase inhibitor used in cancer research. It exhibits IC50 values of 650 nM for EGFR WT and 437 nM for VEGFR2 V916M.
  • Pan-kinase inhibitor
  • Useful in cancer research
  • Shows targeting ability to H1975 cells with VEGFR2 V916M mutation
  • Targets human cancer cell lines such as CRL2064 (wild type), PC9G (EGFR T790M), H3122CR1 (ALK L1196M), and H1975 (VEGFR2 V916M)
Catalog No. 50-003-44178 Supplier Medchemexpress LLC Supplier No. HY11113850MG
CZC-8004 is a pan-kinase inhibitor used in cancer research. It exhibits IC50 values of 650 nM for EGFR WT and 437 nM for VEGFR2 V916M.
  • Pan-kinase inhibitor
  • Useful in cancer research
  • Shows targeting ability to H1975 cells with VEGFR2 V916M mutation
  • Targets human cancer cell lines such as CRL2064 (wild type), PC9G (EGFR T790M), H3122CR1 (ALK L1196M), and H1975 (VEGFR2 V916M)