Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC CZC-8004 | 916603-07-1 | 99.8% | 309.34 | 100 MG
SDP

Catalog No. 5000343380
Encompass_Preferred

CZC-8004 is a pan-kinase inhibitor with IC50 values of 650 nM for EGFR WT and 437 nM for VEGFR2 V916M. It can be used in the study of cancer and shows certain targeting ability to H1975 cells with VEGFR2 V916M mutation in human cancer cell lines.

  • Purity of 99.81%
  • Molecular weight of 309.34
  • Provided as a white to off-white solid
  • Soluble in DMSO at 77.5 mg/mL
  • Powder can be stored at -20°C for 3 years or 4°C for 2 years
  • When in solvent, it can be stored at -80°C for 2 years or -20°C for 1 year

Catalog No. 50-003-43380 Supplier Medchemexpress LLC Supplier No. HY1111381MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CZC-8004 is a pan-kinase inhibitor with IC50 values of 650 nM for EGFR WT and 437 nM for VEGFR2 V916M. It can be used in the study of cancer and shows certain targeting ability to H1975 cells with VEGFR2 V916M mutation in human cancer cell lines.

  • Purity of 99.81%
  • Molecular weight of 309.34
  • Provided as a white to off-white solid
  • Soluble in DMSO at 77.5 mg/mL
  • Powder can be stored at -20°C for 3 years or 4°C for 2 years
  • When in solvent, it can be stored at -80°C for 2 years or -20°C for 1 year

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.