Learn More
Medchemexpress LLC CZC-8004 | 916603-07-1 | 99.81% | 309.34 | 50 MG
SDP
Catalog No. 5000344036
Encompass_Preferred
CZC-8004 is a pan-kinase inhibitor that shows potential for cancer research. It exhibits IC50 values of 650 nM for EGFR WT and 437 nM for VEGFR2 V916M. In vitro, CZC-8004 (1 μM) demonstrates specific targeting capabilities towards H1975 cells that possess the VEGFR2 V916M mutation, with observed activity across a panel of human cancer cell lines.
  • Exhibits IC50 values of 650 nM for EGFR WT and 437 nM for VEGFR2 V916M.
  • Can be used in the study of cancer.
  • Shows certain targeting ability to H1975 cells with VEGFR2 V916M mutation.
  • Observed activity across human cancer cell lines CRL2064 (wild type), PC9G (EGFR T790M), H3122CR1 (ALK L1196M), and H1975 (VEGFR2 V916M).
Catalog No. 50-003-44036 Supplier Medchemexpress LLC Supplier No. HY11113810MM/1ML
CZC-8004 is a pan-kinase inhibitor that shows potential for cancer research. It exhibits IC50 values of 650 nM for EGFR WT and 437 nM for VEGFR2 V916M. In vitro, CZC-8004 (1 μM) demonstrates specific targeting capabilities towards H1975 cells that possess the VEGFR2 V916M mutation, with observed activity across a panel of human cancer cell lines.
  • Exhibits IC50 values of 650 nM for EGFR WT and 437 nM for VEGFR2 V916M.
  • Can be used in the study of cancer.
  • Shows certain targeting ability to H1975 cells with VEGFR2 V916M mutation.
  • Observed activity across human cancer cell lines CRL2064 (wild type), PC9G (EGFR T790M), H3122CR1 (ALK L1196M), and H1975 (VEGFR2 V916M).