Learn More
AB Sciex LLC CZE Rapid Charge Variant Analysis Kit
Catalog No. NC1789025
Encompass
This kit provides all of the necessary reagents to analyze a molecule’s charge variants based on its mobility. This method provides a fast, powerful separation used to quantify charge variants with a buffer that serves as both a separation matrix and dynamic coating for a bare fused silica capillary. In addition, no sample dilution buffer is required; simply dilute the sample in water and you are ready for your separation. CZE Rapid Charge Variant Analysis Kit P/N C44790 includes CZE Rapid Charge Variant Separation Buffer (Qty: 125 mL); Acid Wash/Regenerating Solution (Qty: 100 mL); CE Grade Water (H2O) (Qty: 140 mL); and Protein Test Mix (Qty: 1 vial).
Catalog No. NC1789025 Supplier AB Sciex LLC Supplier No. C44790
This kit provides all of the necessary reagents to analyze a molecule’s charge variants based on its mobility. This method provides a fast, powerful separation used to quantify charge variants with a buffer that serves as both a separation matrix and dynamic coating for a bare fused silica capillary. In addition, no sample dilution buffer is required; simply dilute the sample in water and you are ready for your separation. CZE Rapid Charge Variant Analysis Kit P/N C44790 includes CZE Rapid Charge Variant Separation Buffer (Qty: 125 mL); Acid Wash/Regenerating Solution (Qty: 100 mL); CE Grade Water (H2O) (Qty: 140 mL); and Protein Test Mix (Qty: 1 vial).