Learn More
B BRAUN MED INC OEM D-100 DISCOFIX 1WAY STPCOCK RX
Catalog No. NC1708416
Encompass_Preferred
One-way with female luer lock port and SPIN-LOCK connector, port covers. DEHP-free, Latex-free. Priming volume: 0.20 mL. 100/case
** Please note that one of the following licenses is required in order to process an order:
¿Board of Pharmacy
¿Medical Director
¿Drug/Wholesale Distributor
This item is not returnable. View return policy
Catalog No. NC1708416 Supplier B BRAUN MED INC OEM Supplier No. 455980
 Restricted Item
Add to Cart
Add to Cart
One-way with female luer lock port and SPIN-LOCK connector, port covers. DEHP-free, Latex-free. Priming volume: 0.20 mL. 100/case
** Please note that one of the following licenses is required in order to process an order:
¿Board of Pharmacy
¿Medical Director
¿Drug/Wholesale Distributor