Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

D 102 96.0+%, TCI America™
SDP

Catalog No. D443050MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D443050MG 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D443050MG Supplier TCI America Supplier No. D443050MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 652145-28-3
Molecular Formula C37H30N2O3S2
Molecular Weight (g/mol) 614.778
InChI Key XGMCROHUTRXETK-VQNDASPWSA-N
Synonym 2-[5-[[4-[4-(2,2-Diphenylvinyl)phenyl]-1,2,3,3a,4,8b-hexahydrocyclopenta[b]indol-7-yl]methylene]-4-oxo-2-thioxothiazolidin-3-yl]acetic Acid
PubChem CID 57937215
IUPAC Name 2-[(5Z)-5-[[4-[4-(2,2-diphenylethenyl)phenyl]-2,3,3a,8b-tetrahydro-1H-cyclopenta[b]indol-7-yl]methylidene]-4-oxo-2-sulfanylidene-1,3-thiazolidin-3-yl]acetic acid
SMILES C1CC2C(C1)N(C3=C2C=C(C=C3)C=C4C(=O)N(C(=S)S4)CC(=O)O)C5=CC=C(C=C5)C=C(C6=CC=CC=C6)C7=CC=CC=C7

Specifications

Quantity 50 mg
Formula Weight 614.78
Percent Purity ≥96.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material D 102
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.