Learn More
Selleck Chemical LLC D(-)-Arabinose S5134-25mg
SDP
Catalog No. 503109064
Encompass_Preferred
D-Arabinose is a reducing sugar useful as a substrate in analysis of arabinose isomerases and synthesis of D-erythroascorbic acid in yeast
Catalog No. 50-310-9064 Supplier Selleck Chemical LLC Supplier No. S513425MG
D-Arabinose is a reducing sugar useful as a substrate in analysis of arabinose isomerases and synthesis of D-erythroascorbic acid in yeast