Learn More
Selleck Chemical LLC D-()-Melezitose S5142-25mg
SDP
Catalog No. 503110771
Encompass_Preferred
Melezitose (D-Melezitose ( )-Melezitose D-melicitose ( )-melicitose melicitose) is a nonreducing trisaccharide sugar that is produced by many plant sap eating insects
Catalog No. 50-311-0771 Supplier Selleck Chemical LLC Supplier No. S514225MG
Melezitose (D-Melezitose ( )-Melezitose D-melicitose ( )-melicitose melicitose) is a nonreducing trisaccharide sugar that is produced by many plant sap eating insects