Learn More
Selleck Chemical LLC D- S9349-25MG
SDP
Catalog No. 5000023748
Encompass_Preferred
Raffinose is a trisaccharide composed of galactose glucose and fructose
Catalog No. 50-000-23748 Supplier Selleck Chemical LLC Supplier No. S934925MG
Raffinose is a trisaccharide composed of galactose glucose and fructose