Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

D 131 98.0+%, TCI America™
SDP

Catalog No. D443150MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D443150MG 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D443150MG Supplier TCI America Supplier No. D443150MG
Add to Cart
Add to Cart

Chemical Identifiers

CAS 652145-29-4
Molecular Formula C35H28N2O2
Molecular Weight (g/mol) 508.621
InChI Key GOTRYMLNXIJMCB-UHFFFAOYSA-N
Synonym 2-Cyano-3-[4-[4-(2,2-diphenylvinyl)phenyl]-1,2,3,3a,4,8b-hexahydrocyclopenta[b]indol-7-yl]acrylic Acid
PubChem CID 66974337
IUPAC Name 2-cyano-3-[4-[4-(2,2-diphenylethenyl)phenyl]-2,3,3a,8b-tetrahydro-1H-cyclopenta[b]indol-7-yl]prop-2-enoic acid
SMILES C1CC2C(C1)N(C3=C2C=C(C=C3)C=C(C#N)C(=O)O)C4=CC=C(C=C4)C=C(C5=CC=CC=C5)C6=CC=CC=C6

Specifications

Color Red-Yellow
Quantity 50 mg
Formula Weight 508.62
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material D 131
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.