Learn More
Andwin Scientific D-16 ADAPTER FOR DEN-1
SDP
Catalog No. NC0003851
Encompass_Preferred
The D16 Adaptor is for use with DEN-1 Densitometers, which are made to fit only tubes with an outer diameter of 18mm. With this adaptor, the DEN-1 Densitometers can now be utilized with 16mm tubes.
Catalog No. NC0003851 Supplier Andwin Scientific Supplier No. D16
Add to Cart
Edge
Add to Cart
The D16 Adaptor is for use with DEN-1 Densitometers, which are made to fit only tubes with an outer diameter of 18mm. With this adaptor, the DEN-1 Densitometers can now be utilized with 16mm tubes.