Learn More
Medchemexpress LLC D-2-amino-5-phosphonovaleric acid | 79055-68-8 | MFCD00078839 | >95.0% | 197.13 g/mol | C5H12NO5P | 1 ML
SDP
Catalog No. 5000214360
Encompass_Preferred
D-AP5 (D-APV; D-2-amino-5-phosphonovaleric acid) is a selective, competitive NMDA receptor antagonist (Kd ≈ 1.4 μM) that inhibits the glutamate binding site of NMDA receptors. It is used as a pharmacological tool in neuroscience to block NMDA receptor-mediated responses. The compound is supplied as a solid with manufacturer guidance for preparing defined stock solutions and specified storage conditions.
  • Selective, competitive NMDA receptor antagonist.
  • Blocks the glutamate binding site of NMDA receptors.
  • Useful for studying NMDA-dependent synaptic transmission and plasticity.
  • Supplied as a powder with guidance to prepare defined stock solutions.
  • Storage guidance: powder -20°C; solutions -80°C (long-term) or -20°C (short-term).
  • Defined chemical identifiers: CAS 79055-68-8; molecular weight 197.13 g/mol.
Catalog No. 50-002-14360 Supplier Medchemexpress LLC Supplier No. HY100714A10MM 1ML
D-AP5 (D-APV; D-2-amino-5-phosphonovaleric acid) is a selective, competitive NMDA receptor antagonist (Kd ≈ 1.4 μM) that inhibits the glutamate binding site of NMDA receptors. It is used as a pharmacological tool in neuroscience to block NMDA receptor-mediated responses. The compound is supplied as a solid with manufacturer guidance for preparing defined stock solutions and specified storage conditions.
  • Selective, competitive NMDA receptor antagonist.
  • Blocks the glutamate binding site of NMDA receptors.
  • Useful for studying NMDA-dependent synaptic transmission and plasticity.
  • Supplied as a powder with guidance to prepare defined stock solutions.
  • Storage guidance: powder -20°C; solutions -80°C (long-term) or -20°C (short-term).
  • Defined chemical identifiers: CAS 79055-68-8; molecular weight 197.13 g/mol.