Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC D-2-amino-5-phosphonovaleric acid | 79055-68-8 | MFCD00078839 | 99.9% | 197.13 g/mol | C5H12NO5P | 1 MG
SDP

Catalog No. 5000214538
Encompass_Preferred

D-AP5 is a selective, competitive antagonist of NMDA-type ionotropic glutamate receptors. It binds the glutamate recognition site to inhibit NMDA receptor-mediated synaptic transmission and is widely used in neuroscience research for studying excitatory signaling and synaptic plasticity.

  • Selective, competitive NMDA receptor antagonist.
  • Suitable for in vitro and in vivo studies of NMDA-mediated signaling.
  • Solubility: water soluble (~2 mg/mL with ultrasonic); poor solubility in DMSO and ethanol.
  • Storage: powder stable at -20°C for up to 3 years; store solutions at -80°C for long term.
  • Includes molecular and identifier data to ensure accurate compound selection.

Catalog No. 50-002-14538 Supplier Medchemexpress LLC Supplier No. HY100714A1MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

D-AP5 is a selective, competitive antagonist of NMDA-type ionotropic glutamate receptors. It binds the glutamate recognition site to inhibit NMDA receptor-mediated synaptic transmission and is widely used in neuroscience research for studying excitatory signaling and synaptic plasticity.

  • Selective, competitive NMDA receptor antagonist.
  • Suitable for in vitro and in vivo studies of NMDA-mediated signaling.
  • Solubility: water soluble (~2 mg/mL with ultrasonic); poor solubility in DMSO and ethanol.
  • Storage: powder stable at -20°C for up to 3 years; store solutions at -80°C for long term.
  • Includes molecular and identifier data to ensure accurate compound selection.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.