Learn More
Enzo Life Sciences D-AP5, (5mg), CAS Number: 79055-68-8
SDP
Supplier:  Enzo Life Sciences BMLEA1040005
Encompass_Preferred
NMDA receptor antagonist. Active form of AP5. Potent NMDA receptor antagonist. Alternative Name: D(-)-2-Amino-5-phosphopentanoic acid. Formula: C5H12NO5P. MW: 197.1. CAS Number: 79055-68-8. Purity: ≥99% (HPLC). Appearance: White to off-white solid. Solubility: Soluble in water (acidify for better solubility). Use/Stability: Store, as supplied, at room temperature for up to 1 year. Store solutions at -20°C for up to 3 months. Long Term Storage: Ambient
Catalog No. NC1495262