Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

D-2-Aminoadipic Acid 95.0+%, TCI America™
SDP

Catalog No. A21531G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A21531G 1 g
A21535G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A21531G Supplier TCI America Supplier No. A21531G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 7620-28-2
Molecular Formula C6H11NO4
Molecular Weight (g/mol) 161.157
MDL Number MFCD00063118
InChI Key OYIFNHCXNCRBQI-SCSAIBSYSA-N
Synonym d-2-aminoadipic acid, r-2-aminohexanedioic acid, 2r-2-aminohexanedioic acid, d-a-aminoadipic acid, d-, a-aminoadipic acid, r-2-aminoadipic acid, r-2-amino-hexanedioic acid, hexanedioic acid, 2-amino-,--and +, hexanedioic acid, 2-amino-, 2r, d-homoglutamic acid
PubChem CID 165627
ChEBI CHEBI:37025
IUPAC Name (2R)-2-aminohexanedioic acid
SMILES C(CC(C(=O)O)N)CC(=O)O

Specifications

Melting Point 187°C
Color White-Yellow
Quantity 1 g
Formula Weight 161.16
Percent Purity ≥95.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material D-2-Aminoadipic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.