Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC D-2-aminoglutarimide (hydrochloride) | 1801140-47-5 | MFCD15474942 | 95.0% | 164.59 g/mol | C5H9ClN2O2 | 250 MG
SDP

Catalog No. 5000208922
Encompass_Preferred

D-2-aminoglutarimide hydrochloride is a synthetic intermediate used in the synthesis of hydantoins, including compounds reported to have anti-angiogenic and cancer cell differentiation-inducing activities. It is supplied as a white to off-white solid with typical analytical data (molecular formula C5H9ClN2O2, molecular weight 164.59 g/mol) and is intended for research use in synthetic and medicinal chemistry.

  • Synthetic intermediate for hydantoin synthesis.
  • Reported applications in anti-angiogenic and cancer cell differentiation research.
  • Appearance: white to off-white solid.
  • CAS number: 1801140-47-5.
  • Typical purity: 95.0% (manufacturer COA).
  • Supplied in small research quantities suitable for laboratory synthesis.

Catalog No. 50-002-08922 Supplier Medchemexpress LLC Supplier No. 250MGHYW088629
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

D-2-aminoglutarimide hydrochloride is a synthetic intermediate used in the synthesis of hydantoins, including compounds reported to have anti-angiogenic and cancer cell differentiation-inducing activities. It is supplied as a white to off-white solid with typical analytical data (molecular formula C5H9ClN2O2, molecular weight 164.59 g/mol) and is intended for research use in synthetic and medicinal chemistry.

  • Synthetic intermediate for hydantoin synthesis.
  • Reported applications in anti-angiogenic and cancer cell differentiation research.
  • Appearance: white to off-white solid.
  • CAS number: 1801140-47-5.
  • Typical purity: 95.0% (manufacturer COA).
  • Supplied in small research quantities suitable for laboratory synthesis.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.