Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

D-2-Methyl-1-butanol, Rot.-4.5°

Catalog No. ICN220870 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN220870 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN220870 Supplier MP Biomedicals, Inc Supplier No. 0522087080

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1565-80-6
Molecular Formula C5H12O
Molecular Weight (g/mol) 88.15
InChI Key QPRQEDXDYOZYLA-YFKPBYRVSA-N
Synonym s---2-methyl-1-butanol, s-2-methyl-1-butanol, s-2-methylbutan-1-ol, 1-butanol, 2-methyl-, 2s, 2s-2-methylbutan-1-ol, unii-ii2qjb35ic, 1-butanol, 2-methyl-, s, 2s-2-methyl-1-butanol, s---2-methylbutanol, 2-methyl-1-butanol,-
PubChem CID 2723602
ChEBI CHEBI:50625
IUPAC Name (2S)-2-methylbutan-1-ol
SMILES CCC(C)CO

Specifications

Quantity 100 g
Formula Weight 88.15
Chemical Name or Material D-2-Methyl-1-Butanol, Rot.-4.5°
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.