Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

D-3-Bromocamphor-10-sulfonic acid monohydrate, 99%, Thermo Scientific™

Catalog No. AAA1679106
Change view
Click to view available options
Quantity:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1679106 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAA1679106 Supplier Thermo Scientific Chemicals Supplier No. A1679106

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 209736-59-4
Molecular Formula C10H14BrO4S
MDL Number MFCD00167223,MFCD00167452
Quantity 5g
UN Number UN1759
InChI Key XUJHKPSBHDQIOD-BRDIYROLSA-M
SMILES CC1(C)[C@@H]2CC[C@@]1(CS([O-])(=O)=O)C(=O)[C@@H]2Br
IUPAC Name [(1S,3R,4S)-3-bromo-7,7-dimethyl-2-oxobicyclo[2.2.1]heptan-1-yl]methanesulfonate
Molecular Weight (g/mol) 310.18
PubChem CID 126963546
Formula Weight 329.22 (311.20 Anhydrous)
Percent Purity 99%
Chemical Name or Material D-3-Bromocamphor-10-sulfonic acid monohydrate
Show More Show Less
Hazard Category Danger
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLIDS, N.O.S.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.