Learn More
Selleck Chemical LLC D-3-Phenyllactic acid
SDP
Catalog No. 5000003762
Encompass_Preferred
D-3-Phenyllactic acid is a chiral building block that is used in preparation of statine
Catalog No. 50-000-03762 Supplier Selleck Chemical LLC Supplier No. S632625MG
Add to Cart
Add to Cart
D-3-Phenyllactic acid is a chiral building block that is used in preparation of statine