Learn More
Apexbio Technology LLC D-3-Phenyllactic acid 7326-19-4 100mg
SDP
Catalog No. 5000090465
Encompass_Preferred
D-3-Phenyllactic acid (CAS 7326-19-4) is a small-molecule inhibitor targeting specific Gram-positive and Gram-negative bacterial strains It is designed to disrupt microbial cell membrane integrity and interfere with related cellular metabolic pathways thereby inhibiting bacterial growth D-3-Phenyllactic acid exerts its biological activity primarily through disruption of microbial membranes and interference with cellular metabolism In in vitro studies D-3-Phenyllactic acid demonstrates inhibitory activity with reported IC50 values typically ranging between 10 50 g/mL depending on the target microorganism Based on these pharmacological properties D-3-Phenyllactic acid holds research potential in applications related to pathogen control in food safety and human health
Catalog No. 50-000-90465 Supplier Apexbio Technology LLC Supplier No. C7007100
D-3-Phenyllactic acid (CAS 7326-19-4) is a small-molecule inhibitor targeting specific Gram-positive and Gram-negative bacterial strains It is designed to disrupt microbial cell membrane integrity and interfere with related cellular metabolic pathways thereby inhibiting bacterial growth D-3-Phenyllactic acid exerts its biological activity primarily through disruption of microbial membranes and interference with cellular metabolism In in vitro studies D-3-Phenyllactic acid demonstrates inhibitory activity with reported IC50 values typically ranging between 10 50 g/mL depending on the target microorganism Based on these pharmacological properties D-3-Phenyllactic acid holds research potential in applications related to pathogen control in food safety and human health