Learn More
Medchemexpress LLC D-3263 hydrochloride | 1008763-54-9 | 98.12% | 409.95 | 100 MG
SDP
Catalog No. 5000394208
Encompass_Preferred
D-3263 hydrochloride is an enteric-coated, orally bioavailable transient receptor potential melastatin member 8 (TRPM8) agonist. It binds to and activates TRPM8, which may lead to an increase in calcium and sodium entry, disruption of calcium and sodium homeostasis, and induction of cell death in TRPM8-expressing tumor cells. D-3263 hydrochloride may also decrease dihydrotestosterone (DHT) levels, potentially contributing to its inhibitory effects on prostate cancer and benign prostatic hyperplasia (BPH).
  • Binds to and activates TRPM8.
  • Increases calcium and sodium entry into cells.
  • Disrupts calcium and sodium homeostasis.
  • Induces cell death in TRPM8-expressing tumor cells.
  • Decreases dihydrotestosterone (DHT) levels.
  • Inhibits prostate cancer and benign prostatic hyperplasia (BPH).
Catalog No. 50-003-94208 Supplier Medchemexpress LLC Supplier No. HY16162A100MG
D-3263 hydrochloride is an enteric-coated, orally bioavailable transient receptor potential melastatin member 8 (TRPM8) agonist. It binds to and activates TRPM8, which may lead to an increase in calcium and sodium entry, disruption of calcium and sodium homeostasis, and induction of cell death in TRPM8-expressing tumor cells. D-3263 hydrochloride may also decrease dihydrotestosterone (DHT) levels, potentially contributing to its inhibitory effects on prostate cancer and benign prostatic hyperplasia (BPH).
  • Binds to and activates TRPM8.
  • Increases calcium and sodium entry into cells.
  • Disrupts calcium and sodium homeostasis.
  • Induces cell death in TRPM8-expressing tumor cells.
  • Decreases dihydrotestosterone (DHT) levels.
  • Inhibits prostate cancer and benign prostatic hyperplasia (BPH).