Learn More
Medchemexpress LLC D-3263 hydrochloride | 1008763-54-9 | MFCD22665741 | 98.0% | 409.95 | C21H32ClN3O3 | 10 MG
SDP
Catalog No. 5000288607
Encompass_Preferred
D-3263 hydrochloride is an enteric-coated, orally bioavailable agonist of transient receptor potential melastatin member 8 (TRPM8). It binds and activates TRPM8, modulates calcium and sodium flux, and has been investigated for potential antineoplastic effects and modulation of androgen-related pathways.
  • Orally bioavailable TRPM8 agonist suitable for in vivo studies.
  • Enteric-coated formulation for improved gastrointestinal stability.
  • High purity (98.0%) supports reproducible experimental results.
  • Soluble in DMSO and formulatable for common in vivo vehicles.
  • Stable when stored sealed at 4°C; solvent stability at -80°C for extended storage.
Catalog No. 50-002-88607 Supplier Medchemexpress LLC Supplier No. HY16162A10MG
D-3263 hydrochloride is an enteric-coated, orally bioavailable agonist of transient receptor potential melastatin member 8 (TRPM8). It binds and activates TRPM8, modulates calcium and sodium flux, and has been investigated for potential antineoplastic effects and modulation of androgen-related pathways.
  • Orally bioavailable TRPM8 agonist suitable for in vivo studies.
  • Enteric-coated formulation for improved gastrointestinal stability.
  • High purity (98.0%) supports reproducible experimental results.
  • Soluble in DMSO and formulatable for common in vivo vehicles.
  • Stable when stored sealed at 4°C; solvent stability at -80°C for extended storage.