Learn More
TARGETMOL CHEMICALS INC D-3263 HYDROCHLORIDE 5MG
SDP
Catalog No. 502577456
Encompass_Preferred
Also available in 1mg 10mg 25mg 50mg and bulk. Please contact Fisher for quotes. D-3263 hydrochloride (D3263 HCl salt) is enteric-coated orally bioavailable transient receptor potential melatonin member 8 (TRPM8) agonist. purity: 99%
Catalog No. 50-257-7456 Supplier TARGETMOL CHEMICALS INC Supplier No. T10929L5MG
Also available in 1mg 10mg 25mg 50mg and bulk. Please contact Fisher for quotes. D-3263 hydrochloride (D3263 HCl salt) is enteric-coated orally bioavailable transient receptor potential melatonin member 8 (TRPM8) agonist. purity: 99%