Learn More
Medchemexpress LLC HY-16162A 5mg Medchemexpress, D-3263 (hydrochloride) CAS:1008763-54-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY16162A5MG
Encompass_Preferred
Medchemexpress, HY-16162A 5mg D-3263 (hydrochloride) CAS:1008763-54-9 D-3263 hydrochloride is an enteric-coated, orally bioavailable (transient receptor potential melastatin member 8) TRPM8 agonist. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1871