Learn More
Selleck Chemical LLC D 4476 10mg 301836-43-1
SDP
Supplier:  Selleck Chemical LLC S764210MG
Encompass_Preferred
D 4476 is a potent, selective, and cell-permeant CK1 (casein kinase 1) inhibitor with IC50 of 200 nM and 300 nM in a cell-free assay for CK1 from Schizosaccharomyces pombe and CK1?, respectively. Also acts as an ALK5 inhibitor with IC50 of 500 nM. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-5063