Learn More
Selleck Chemical LLC D 4476
SDP
Catalog No. 5000000219
Encompass_Preferred
D 4476 is a potent selective and cell-permeant CK1 (casein kinase 1) inhibitor with IC50 of 200 nM and 300 nM in a cell-free assay for CK1 from Schizosaccharomyces pombe and CK1 respectively Also acts as an ALK5 inhibitor with IC50 of 500 nM
Catalog No. 50-000-00219 Supplier Selleck Chemical LLC Supplier No. S764210MM/1ML
D 4476 is a potent selective and cell-permeant CK1 (casein kinase 1) inhibitor with IC50 of 200 nM and 300 nM in a cell-free assay for CK1 from Schizosaccharomyces pombe and CK1 respectively Also acts as an ALK5 inhibitor with IC50 of 500 nM