Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ D.A.R.T.'S™ (Disposable Automation Research Tips)

Catalog No. 14387664 Shop All Thermo Scientific Products
Change view
Click to view available options
Volume (Metric):
100 μL
Sterility:
Non-sterile
Filtered:
Non-filtered
Packaging:
20 x 384 tips
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) Sterility Filtered Packaging
14-387-664 100 μL Non-sterile Non-filtered 20 x 384 tips
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 14-387-664 Supplier Thermo Scientific™ Supplier No. 5326

missing translation for 'validMainframeMsg'

Achieve ultimate performance in automated pipetting with these tips--superior quality with the flexibility to choose tips for any application.

Perform pipetting more effectively with Thermo Scientific™ Matrix™ SerialMate™ Pipette Tips, for use with the Matrix SerialMate™ Pipetter. Unlike conventional automation tips, D.A.R.T.'s™ are packaged in disposable rigid magazines and seal onto the pipetting heads by being pressed against a silicone pad. This definitive seal minimizes volume displacement, ensures tip straightness and enables consistent dispensing for improved well access. A small internal orifice provides small droplet formation resulting in optimal accuracy and precision.

Features:

  • Stepped-body design minimizes volume displacement and ensures tip straightness for better well access
  • Highly polished molds and cores ensure low liquid retention
  • Small internal tip orifice provides better small volume droplet formation
  • Sterile, sterile-filtered, wide-bore, and extended-length tip options are available
  • Tips are thoroughly inspected for proper formation and straightness
  • Unique face seal design ensures consistent tip-to-tip height for precise low volume dispensing
  • Entire assembly can be quickly removed and replaced eliminating extra handling of the tips, which can cause damage or contamination

Includes: 20 magazines per case

Compatible with: Thermo Scientific™ Matrix™ Hydra™ DT, Matrix™ PlateMate™ 2x3 and PlateMate™ Plus Systems.

TRUSTED_SUSTAINABILITY

Specifications

Volume (Metric) 100 μL
Filtered Non-filtered
Sterility Non-sterile
Color Natural
Autoclavable Autoclavable
Length (English) Standard
Certifications/Compliance ISO 9001 and ISO 13485
Clean Claims Free of RNase, DNase, DNA, ATP, and endotoxin
Format Rack
Tip Style Automation
Packaging 20 x 384 tips
Quantity 7680 tips
Disposable Single-use
For Use With (Equipment) Thermo Scientific Matrix Hydra DT, Thermo Scientific PlateMate Plus, Thermo Scientific Versette
Product Line Matrix
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.