Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC D-a-tert-Butyl-Gly-OH | 26782-71-8 | 131.17 | 100 G
SDP

Catalog No. 5000447866
Encompass_Preferred

D-a-tert-Butyl-Gly-OH is a leucine derivative, categorized as an amino acid derivative. These compounds are utilized as ergogenic supplements, impacting anabolic hormone secretion, providing fuel during exercise, improving mental performance under stress, and aiding in the prevention of exercise-induced muscle damage. They are widely recognized as beneficial ergogenic dietary substances.

  • Leucine derivative
  • Functions as an ergogenic supplement
  • Influences anabolic hormone secretion
  • Supports fuel supply during physical activity
  • Enhances mental performance during stress-related tasks
  • Assists in preventing exercise-induced muscle damage
  • Purity: ≥98.0%
  • Molecular weight: 131.17
  • Formula: C6H13NO2
  • Appearance: Solid
  • Color: White to off-white
  • Solubility in H2O: 33.33 mg/mL (254.10 mM)

Catalog No. 50-004-47866 Supplier Medchemexpress LLC Supplier No. HYW001954100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

D-a-tert-Butyl-Gly-OH is a leucine derivative, categorized as an amino acid derivative. These compounds are utilized as ergogenic supplements, impacting anabolic hormone secretion, providing fuel during exercise, improving mental performance under stress, and aiding in the prevention of exercise-induced muscle damage. They are widely recognized as beneficial ergogenic dietary substances.

  • Leucine derivative
  • Functions as an ergogenic supplement
  • Influences anabolic hormone secretion
  • Supports fuel supply during physical activity
  • Enhances mental performance during stress-related tasks
  • Assists in preventing exercise-induced muscle damage
  • Purity: ≥98.0%
  • Molecular weight: 131.17
  • Formula: C6H13NO2
  • Appearance: Solid
  • Color: White to off-white
  • Solubility in H2O: 33.33 mg/mL (254.10 mM)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.