Learn More
Cayman Chemical D-a-Tocopheryl QuInon 10mg
SDP
Catalog No. 502913299
Encompass_Preferred
An oxidative metabolite of vitamin E; has antioxidant activity in ORAC and radical trapping assays at 25 µM; inhibits (1S,3R)-RSL3-induced ferroptosis in Q7 cells (EC50 = 0.233 µM); decreases serum MDA and blood glucose levels and restores the number and diversity of intestinal microflora in a rat model of high cholesterol and cholate diet-induced NASH at 20 mg/kg
Catalog No. 50-291-3299 Supplier Cayman Chemical Supplier No. 3536510 MG
An oxidative metabolite of vitamin E; has antioxidant activity in ORAC and radical trapping assays at 25 µM; inhibits (1S,3R)-RSL3-induced ferroptosis in Q7 cells (EC50 = 0.233 µM); decreases serum MDA and blood glucose levels and restores the number and diversity of intestinal microflora in a rat model of high cholesterol and cholate diet-induced NASH at 20 mg/kg