Learn More
TARGETMOL CHEMICALS INC D-Ala-Lys-AMCA hydrochloride
SDP
Catalog No. 502878581
Encompass_Preferred
Also available in 1 mL, 5 mg, 10 mg, 25 mg, 50 mg and bulk. Please contact Fisher for quotes. D-Ala-Lys-AMCA hydrochloride, a known substrate of proton-coupled oligopeptide transporter 1 (PEPT1), emits blue fluorescence and may be transported into Caco-2 cells and liver cancer cells. Purity 98.83%
Catalog No. 50-287-8581 Supplier TARGETMOL CHEMICALS INC Supplier No. T189571MG
Also available in 1 mL, 5 mg, 10 mg, 25 mg, 50 mg and bulk. Please contact Fisher for quotes. D-Ala-Lys-AMCA hydrochloride, a known substrate of proton-coupled oligopeptide transporter 1 (PEPT1), emits blue fluorescence and may be transported into Caco-2 cells and liver cancer cells. Purity 98.83%