Learn More
Bachem (D-Ala2,D-Leu5)-Enkephalin Acetate
SDP
Supplier:  Bachem 4001508.0025
Encompass_Preferred
(D-Ala2,D-Leu5)-Enkephalin Acetate, H-Tyr-D-Ala-Gly-Phe-D-Leu-OH, 63631-40-3, C29H39N5O7, Mr 569.66, 25mg. DADLE, [DAla2, DLeu5] Enkephalin. DADLE has generally served as the putative, prototypical d-opioid receptor ligand. DADLE is more potent and selective than Leu-enkephalin or Met-enkephalin in the MVD versus GPI bioassay systems and more potent and approximately as selective as Leu-enkephalin in binding assays. It shows roughly the same antinociceptive activity as b-endorphin.
Catalog No. 50-259-658