Learn More
Medchemexpress LLC [D-Ala2, D-Leu5]-enkephalin | 63631-40-3 | 99.9% | 569.65 | C29H39N5O7 | 100 MG
SDP
Catalog No. 5000242390
Encompass_Preferred
DADLE ([D-Ala2, D-Leu5]-enkephalin) is a synthetic enkephalin-family peptide that acts as a selective delta-opioid receptor agonist. Supplied for research use only, it is used in neuropharmacology, receptor signaling assays, and studies of neuroprotection.
  • Acts as a selective delta-opioid receptor agonist.
  • Synthetic enkephalin peptide with sequence modifications for activity.
  • Molecular formula C29H39N5O7, molecular weight 569.65 g/mol.
  • High purity (99.9%).
  • Available as solid in multiple sizes, including 100 MG, and as 10 mM solution in DMSO.
  • For research use only; not for human or clinical use.
Catalog No. 50-002-42390 Supplier Medchemexpress LLC Supplier No. HY105343100MG
DADLE ([D-Ala2, D-Leu5]-enkephalin) is a synthetic enkephalin-family peptide that acts as a selective delta-opioid receptor agonist. Supplied for research use only, it is used in neuropharmacology, receptor signaling assays, and studies of neuroprotection.
  • Acts as a selective delta-opioid receptor agonist.
  • Synthetic enkephalin peptide with sequence modifications for activity.
  • Molecular formula C29H39N5O7, molecular weight 569.65 g/mol.
  • High purity (99.9%).
  • Available as solid in multiple sizes, including 100 MG, and as 10 mM solution in DMSO.
  • For research use only; not for human or clinical use.