Learn More
Medchemexpress LLC [D-Ala2, D-Leu5]-enkephalin | 63631-40-3 | 99.9% | 569.65 g/mol | C29H39N5O7 | 10 MG
SDP
Catalog No. 5000244316
Encompass_Preferred
BW-180C ([D-Ala2, D-Leu5]-enkephalin, DADLE) is a synthetic enkephalin peptide that functions as a δ-opioid receptor (DOR) agonist. It is intended for research use in pharmacology and neuroscience to study opioid receptor signaling and neuroprotective effects. The material is supplied as a high-purity solid and as pre-made DMSO solutions, with recommended low-temperature storage for long-term stability.
  • Selective δ-opioid receptor agonist for receptor pharmacology studies.
  • High purity suitable for biochemical and cell-based assays.
  • Stable as powder and in DMSO under recommended storage conditions.
  • Available in small research quantities and solution formats.
  • Analytical documentation provided for quality verification.
Catalog No. 50-002-44316 Supplier Medchemexpress LLC Supplier No. HY10534310MG
BW-180C ([D-Ala2, D-Leu5]-enkephalin, DADLE) is a synthetic enkephalin peptide that functions as a δ-opioid receptor (DOR) agonist. It is intended for research use in pharmacology and neuroscience to study opioid receptor signaling and neuroprotective effects. The material is supplied as a high-purity solid and as pre-made DMSO solutions, with recommended low-temperature storage for long-term stability.
  • Selective δ-opioid receptor agonist for receptor pharmacology studies.
  • High purity suitable for biochemical and cell-based assays.
  • Stable as powder and in DMSO under recommended storage conditions.
  • Available in small research quantities and solution formats.
  • Analytical documentation provided for quality verification.