Learn More
Medchemexpress LLC [D-Ala2, D-Leu5]-enkephalin | 63631-40-3 | MFCD00076411 | 99.9% | 569.65 | C29H39N5O7 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY10534310MM 1ML
Encompass_Preferred
BW-180C (DADLE; [D-Ala2, D-Leu5]-enkephalin) is a synthetic enkephalin peptide that functions as a selective delta-opioid receptor agonist. It is supplied for research use in neuropharmacology and GPCR signaling studies, and is available as lyophilized solid or as a ready-to-use 10 mM solution in DMSO. Documentation includes datasheet, COA, SDS, and spectral data.
  • Selective delta-opioid receptor agonist useful in receptor pharmacology.
  • Synthetic enkephalin peptide sequence [D-Ala2, D-Leu5]-enkephalin.
  • Supplied as lyophilized solid and 10 mM solution in DMSO (1 mL).
  • High purity suitable for in vitro assays (reported 99.9%).
  • Molecular weight 569.65 and formula C29H39N5O7.
  • Includes datasheet, certificate of analysis, SDS, and spectral data for quality assessment.
Catalog No. 50-002-43918