Learn More
Medchemexpress LLC [D-Ala2]-GIP (human) | 444073-04-5 | 98.97% | 4983.60 | 25 MG
SDP
Catalog No. 5000398970
Encompass_Preferred
[D-Ala2]-GIP (human) is a GIP receptor agonist with an EC50 of 630 pM. It improves glucose tolerance and demonstrates neuroprotective activity in an MPTP-induced Parkinson's disease model. Additionally, it enhances cognitive function and hippocampal synaptic plasticity in obese diabetic rats. This product is suitable for research related to type 2 diabetes, Parkinson's disease, and similar conditions.
  • Acts as a GIP receptor agonist with an EC50 of 630 pM
  • Improves glucose tolerance by stimulating insulin release
  • Demonstrates neuroprotective activity in MPTP-induced Parkinson's disease models
  • Enhances cognitive function and hippocampal synaptic plasticity in obese diabetic rats
  • Attenuates the behavioral phenotype of Parkinson's disease
  • Reverses decline in motor ability and reduces muscle stiffness
  • Inhibits the increase of MDA levels in the brain
Catalog No. 50-003-98970 Supplier Medchemexpress LLC Supplier No. 25MGHYP3247
[D-Ala2]-GIP (human) is a GIP receptor agonist with an EC50 of 630 pM. It improves glucose tolerance and demonstrates neuroprotective activity in an MPTP-induced Parkinson's disease model. Additionally, it enhances cognitive function and hippocampal synaptic plasticity in obese diabetic rats. This product is suitable for research related to type 2 diabetes, Parkinson's disease, and similar conditions.
  • Acts as a GIP receptor agonist with an EC50 of 630 pM
  • Improves glucose tolerance by stimulating insulin release
  • Demonstrates neuroprotective activity in MPTP-induced Parkinson's disease models
  • Enhances cognitive function and hippocampal synaptic plasticity in obese diabetic rats
  • Attenuates the behavioral phenotype of Parkinson's disease
  • Reverses decline in motor ability and reduces muscle stiffness
  • Inhibits the increase of MDA levels in the brain