Learn More
Medchemexpress LLC [D-Ala2]-GIP (human) | 444073-04-5 | 99.0% | 4983.60 | 5 MG
SDP
Catalog No. 5000397301
Encompass_Preferred
[D-Ala2]-GIP (human) is a GIP receptor agonist (EC50 = 630 pM) that improves glucose tolerance and shows neuroprotective activity. It also enhances cognitive function and hippocampal synaptic plasticity, making it suitable for research in type 2 diabetes, Parkinson's disease, and related areas. This product is for research use only.
  • Significantly reduces blood glucose fluctuations by stimulating insulin release.
  • Attenuates the behavioral phenotype of Parkinson's disease.
  • Reverses the decline in motor ability and muscle stiffness.
  • Reduces increased tremor amplitude and inhibits the increase of MDA levels in the brain.
  • Improves cognitive function and hippocampal synaptic plasticity.
Catalog No. 50-003-97301 Supplier Medchemexpress LLC Supplier No. HYP32475MG
[D-Ala2]-GIP (human) is a GIP receptor agonist (EC50 = 630 pM) that improves glucose tolerance and shows neuroprotective activity. It also enhances cognitive function and hippocampal synaptic plasticity, making it suitable for research in type 2 diabetes, Parkinson's disease, and related areas. This product is for research use only.
  • Significantly reduces blood glucose fluctuations by stimulating insulin release.
  • Attenuates the behavioral phenotype of Parkinson's disease.
  • Reverses the decline in motor ability and muscle stiffness.
  • Reduces increased tremor amplitude and inhibits the increase of MDA levels in the brain.
  • Improves cognitive function and hippocampal synaptic plasticity.