Learn More
Phoenix Pharmaceuticals Inc D-ALA2-GIP Rat, GIP Receptor Agonist, Improves Glucose Tolerance, 2 mg
Catalog No. NC1782237
Encompass
D-ALA2-GIP (Rat) is a synthetic peptide designed for research, enhancing glucose tolerance and stimulating insulin release. Its unique D-Alanine modification at position 2 ensures resistance to degradation, prolonging its in vivo effectiveness. This potent GIP receptor agonist is invaluable for studying metabolic processes in rat models, distinguishing it as a superior tool in diabetes research and glucose homeostasis studies.
Catalog No. NC1782237 Supplier Phoenix Pharmaceuticals Inc Supplier No. 02714
D-ALA2-GIP (Rat) is a synthetic peptide designed for research, enhancing glucose tolerance and stimulating insulin release. Its unique D-Alanine modification at position 2 ensures resistance to degradation, prolonging its in vivo effectiveness. This potent GIP receptor agonist is invaluable for studying metabolic processes in rat models, distinguishing it as a superior tool in diabetes research and glucose homeostasis studies.