Learn More
Medchemexpress LLC HY-P0098 5mg Medchemexpress, [D-Ala2]leucine-enkephalin CAS:64963-01-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYP00985MG
Encompass_Preferred
Medchemexpress, HY-P0098 5mg [D-Ala2]leucine-enkephalin CAS:64963-01-5 [D-Ala2]leucine-enkephalin, a delta opioid agonist, is a degradation resistant long-acting Leu-enkephalin. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-4021