Learn More
Medchemexpress LLC [D-Ala2]leucine-enkephalin | 64963-01-5 | 98.6% | 569.65 | 1 ML
SDP
Catalog No. 5000426062
Encompass_Preferred
[D-Ala2]leucine-enkephalin is a delta opioid agonist and a degradation-resistant, long-acting Leu-enkephalin analog. It is used to study the signaling pathway of delta opioid receptors. It is widely distributed in the human body and possesses vital physiological functions.
  • Activates dormant brain cells and facilitates recovery of sequelae from brain damage.
  • Enhances immune system activity, including T lymphocytes, NK cells, and macrophages.
  • Exerts antiviral effects and promotes gamma-interferon secretion.
  • Participates in immune regulation by stimulating endocrine cell secretion, influencing gene regulation and DNA expression.
Catalog No. 50-004-26062 Supplier Medchemexpress LLC Supplier No. HYP009810MM/1ML
[D-Ala2]leucine-enkephalin is a delta opioid agonist and a degradation-resistant, long-acting Leu-enkephalin analog. It is used to study the signaling pathway of delta opioid receptors. It is widely distributed in the human body and possesses vital physiological functions.
  • Activates dormant brain cells and facilitates recovery of sequelae from brain damage.
  • Enhances immune system activity, including T lymphocytes, NK cells, and macrophages.
  • Exerts antiviral effects and promotes gamma-interferon secretion.
  • Participates in immune regulation by stimulating endocrine cell secretion, influencing gene regulation and DNA expression.